ENA458438107 Enamine

2-(pyridin-3-yl)pyridin-3-amine

  • MDL number MFCD15475157
  • Cas Number : 170808-76-1
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-239901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-239901
Availability details for
Quantity
  • Availability
  • Add to Cart