ENA458441183 Enamine

1-(2,3-dimethoxyphenyl)propan-1-amine

  • MDL number MFCD18703470
  • Cas Number : 1270381-49-1
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-242981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-242981
Availability details for
Quantity
  • Availability
  • Add to Cart