ENA458441502 Enamine

N-[1-(1,3-dimethyl-1H-pyrazol-4-yl)ethylidene]hydroxylamine

  • MDL number MFCD20491468
  • Cas Number : 1005583-56-1
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64431
Availability details for
Quantity
  • Availability
  • Add to Cart