ENA458441563 Enamine

N-[1-phenyl-2-(1H-pyrazol-1-yl)ethylidene]hydroxylamine

  • Cas Number : 99205-91-1
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-79653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79653
Availability details for
Quantity
  • Availability
  • Add to Cart