ENA465562126 Enamine

2-(3-methylpyridin-2-yl)ethan-1-ol

  • MDL number MFCD06637708
  • Cas Number : 4723-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-218264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218264
Availability details for
Quantity
  • Availability
  • Add to Cart