ENA465562549 Enamine

4-(propan-2-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD19282029
  • Cas Number : 81569-42-8
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-172212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172212
Availability details for
Quantity
  • Availability
  • Add to Cart