ENA465563640 Enamine

N,N-dimethyl-3-azabicyclo[3.1.0]hexan-6-amine dihydrochloride

  • MDL number MFCD28148308
  • Cas Number : 1909337-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-243821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-243821
Availability details for
Quantity
  • Availability
  • Add to Cart