ENA465564542 Enamine

1-(4-aminophenyl)-2-methylpropan-1-one

  • MDL number MFCD03416574
  • Cas Number : 95249-12-0
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85279

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-85279
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart