ENA465564584 Enamine

heptanenitrile

  • MDL number MFCD00039503
  • Cas Number : 629-08-3
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 58
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99427

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P270,P271,P280,P301+P312,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P311,P312,P330,P332+P313,P337+P313,P362,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-99427
s_phrases_eu 26-36
pictograms GHS02,GHS06
hstatements H226,H302,H312,H315,H319,H331
Availability details for
Quantity
  • Availability
  • Add to Cart