ENA490026568 Enamine

2H-[1,3]dioxolo[4,5-b]pyridin-5-amine

  • MDL number MFCD22071170
  • Cas Number : 1368345-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134540
Availability details for
Quantity
  • Availability
  • Add to Cart