ENA490026625 Enamine

1-(4-chloro-3-hydroxyphenyl)ethan-1-one

  • MDL number MFCD00034670
  • Cas Number : 61124-56-9
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-179326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179326
Availability details for
Quantity
  • Availability
  • Add to Cart