ENA490046798 Enamine

1,3-diethyl 2-methyl-2-(5-oxopyrrolidin-2-yl)propanedioate

  • MDL number MFCD29047195
  • Cas Number : 1909306-00-8
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-227565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227565
Availability details for
Quantity
  • Availability
  • Add to Cart