ENA490046840 Enamine

ethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-2-carboxylate

  • MDL number MFCD11975790
  • Cas Number : 307307-84-2
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-244259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-244259
Availability details for
Quantity
  • Availability
  • Add to Cart