ENA490047113 Enamine

4-amino-N-(1H-imidazol-2-yl)-1-methyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD21221531
  • Cas Number : 1480596-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-247328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-247328
Availability details for
Quantity
  • Availability
  • Add to Cart