ENA490047544 Enamine

6-(trifluoromethyl)isoquinolin-1-amine

  • MDL number MFCD13189943
  • Cas Number : 1196157-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-248629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-248629
Availability details for
Quantity
  • Availability
  • Add to Cart