ENA490047656 Enamine

5-(chloromethyl)-N,N-dimethylpyridin-2-amine hydrochloride

  • MDL number MFCD29047476
  • Cas Number : 1909316-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-248951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-248951
Availability details for
Quantity
  • Availability
  • Add to Cart