ENA497372789 Enamine

tert-butyl N-[2-(aminomethyl)adamantan-2-yl]carbamate

  • Cas Number : 1909337-27-4
  • Molecular Weight : 280.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-246691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-246691
Availability details for
Quantity
  • Availability
  • Add to Cart