ENA497372845 Enamine

2-{[(tert-butoxy)carbonyl]amino}pent-4-ynoic acid

  • MDL number MFCD18207317
  • Cas Number : 61172-66-5
  • Molecular Weight : 213.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-118265

Safety Information

signalword Warning
pcodes P261,P272,P280,P302+P352,P333+P313,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-118265
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart