ENA497373112 Enamine

2-{[(tert-butoxy)carbonyl]amino}-3-cyclohexylpropanoic acid

  • MDL number MFCD03095514
  • Cas Number : 144186-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203491
Availability details for
Quantity
  • Availability
  • Add to Cart