ENA497373283 Enamine

3-(pyrimidin-5-yl)propan-1-amine

  • MDL number MFCD14636533
  • Cas Number : 112104-02-6
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-220345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220345
Availability details for
Quantity
  • Availability
  • Add to Cart