ENA497373305 Enamine

1-[(4-aminophenyl)imino]-1lambda6-thiolan-1-one

  • MDL number MFCD29762617
  • Cas Number : 910780-69-7
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-222385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222385
Availability details for
Quantity
  • Availability
  • Add to Cart