ENA497373338 Enamine

2-cyclopropylcyclohexane-1-carboxylic acid

  • MDL number MFCD24025058
  • Cas Number : 1512893-46-7
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-225887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225887
Availability details for
Quantity
  • Availability
  • Add to Cart