ENA497373683 Enamine

2-(trifluoromethyl)-1,3-benzothiazole-4-carboxylic acid

  • MDL number MFCD28535811
  • Cas Number : 1781508-50-6
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-248698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-248698
Availability details for
Quantity
  • Availability
  • Add to Cart