ENA497373729 Enamine

tert-butyl 3-(piperidin-2-yl)propanoate

  • MDL number MFCD16693003
  • Cas Number : 1249196-03-9
  • Molecular Weight : 213.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-249106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-249106
Availability details for
Quantity
  • Availability
  • Add to Cart