ENA497373773 Enamine

{4-[amino(phenyl)methyl]oxan-4-yl}methanol

  • MDL number MFCD29762681
  • Cas Number : 1909305-33-4
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-249842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-249842
Availability details for
Quantity
  • Availability
  • Add to Cart