ENA497374094 Enamine

(2R,3S)-3-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)-2-methylmorpholine hydrochloride

  • MDL number MFCD29054974
  • Cas Number : 1867912-68-2
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-250963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-250963
Availability details for
Quantity
  • Availability
  • Add to Cart