ENA497374120 Enamine

3-cyclopropyl-3-methoxybutan-2-one

  • MDL number MFCD26022977
  • Cas Number : 1549538-25-1
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-251319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-251319
Availability details for
Quantity
  • Availability
  • Add to Cart