ENA497374282 Enamine

4-chloro-N-(2-ethoxyethyl)aniline hydrochloride

  • MDL number MFCD29055018
  • Cas Number : 1909308-70-8
  • Molecular Weight : 236.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-251790

Safety Information

signalword Warning
pcodes P264,P280,P302+P352,P332+P313,P362
MSDS https://www.enaminestore.com/catalog/EN300-251790
pictograms GHS07
hstatements H315
Availability details for
Quantity
  • Availability
  • Add to Cart