ENA497374522 Enamine

2-cyclopropyl-3-(pyrrolidin-1-yl)propan-1-amine

  • MDL number MFCD29060306
  • Cas Number : 1894757-22-2
  • Molecular Weight : 168.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-252482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252482
Availability details for
Quantity
  • Availability
  • Add to Cart