ENA497374636 Enamine

methyl 4-cyano-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD13178586
  • Cas Number : 40740-43-0
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-252765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252765
Availability details for
Quantity
  • Availability
  • Add to Cart