ENA497374675 Enamine

3-chloro-5-[2-(trimethylsilyl)ethynyl]pyridine

  • MDL number MFCD27949463
  • Cas Number : 329202-21-3
  • Molecular Weight : 209.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-252823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252823
Availability details for
Quantity
  • Availability
  • Add to Cart