ENA497374730 Enamine

5-chloro-1-methyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD16619821
  • Cas Number : 111493-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-252947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252947
Availability details for
Quantity
  • Availability
  • Add to Cart