ENA497374768 Enamine

2-phenylbenzene-1,3-diol

  • MDL number MFCD24713632
  • Cas Number : 3796-74-5
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-253091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253091
Availability details for
Quantity
  • Availability
  • Add to Cart