ENA497374835 Enamine

3-tert-butyl-N-methylcyclobutan-1-amine hydrochloride

  • MDL number MFCD29055148
  • Cas Number : 1955539-89-5
  • Molecular Weight : 177.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-253169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253169
Availability details for
Quantity
  • Availability
  • Add to Cart