ENA497375011 Enamine

(3S)-3-{[(2S)-2-amino-4-methylpentanamido]methyl}-5-methylhexanoic acid hydrochloride

  • MDL number MFCD29055178
  • Cas Number : 1909287-89-3
  • Molecular Weight : 308.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-253564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253564
Availability details for
Quantity
  • Availability
  • Add to Cart