ENA497375084 Enamine

2-(morpholin-4-yl)-2-(1,3-thiazol-4-yl)acetonitrile

  • MDL number MFCD29055187
  • Cas Number : 1602501-52-9
  • Molecular Weight : 209.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-253770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253770
Availability details for
Quantity
  • Availability
  • Add to Cart