ENA497375418 Enamine

cyclopentyl(1,5-dimethyl-1H-pyrazol-3-yl)methanol

  • MDL number MFCD29762797
  • Cas Number : 1698758-27-8
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-254701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-254701
Availability details for
Quantity
  • Availability
  • Add to Cart