ENA497375440 Enamine

1-cyclopentyl-5-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13501681
  • Cas Number : 1306092-80-7
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-254781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-254781
Availability details for
Quantity
  • Availability
  • Add to Cart