ENA497375465 Enamine

3-[3-(trifluoromethyl)phenyl]-1,2-thiazol-4-amine

  • MDL number MFCD21280622
  • Cas Number : 1498589-74-4
  • Molecular Weight : 244.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-254842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-254842
Availability details for
Quantity
  • Availability
  • Add to Cart