ENA497375722 Enamine

2,3,4-trimethylpyrrolidine

  • MDL number MFCD19222298
  • Cas Number : 51308-51-1
  • Molecular Weight : 113.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-255439

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-255439
pictograms GHS02,GHS05,GHS07
hstatements H225,H302,H312,H314,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart