ENA497376305 Enamine

3-bromo-5-methyl-1-benzofuran

  • MDL number MFCD22560092
  • Cas Number : 1388071-39-3
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-256694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-256694
Availability details for
Quantity
  • Availability
  • Add to Cart