ENA497376326 Enamine

5-aminoazepan-2-one dihydrochloride

  • MDL number MFCD29060549
  • Cas Number : 1909319-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-256759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-256759
Availability details for
Quantity
  • Availability
  • Add to Cart