ENA497376543 Enamine

7-amino-1,2-dihydroisoquinolin-1-one hydrochloride

  • MDL number MFCD29762938
  • Cas Number : 1909309-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-257317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-257317
Availability details for
Quantity
  • Availability
  • Add to Cart