ENA497376615 Enamine

N-[4-(aminomethyl)phenyl]-4-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD26459259
  • Cas Number : 1909336-20-4
  • Molecular Weight : 267.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-257450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-257450
Availability details for
Quantity
  • Availability
  • Add to Cart