ENA497376698 Enamine

1-(2,2-dimethylcyclopropyl)propan-1-amine

  • MDL number MFCD29060483
  • Cas Number : 1702089-50-6
  • Molecular Weight : 127.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-255792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-255792
Availability details for
Quantity
  • Availability
  • Add to Cart