ENA497377217 Enamine

4-[(pyrrolidin-3-yl)methyl]piperazin-2-one dihydrochloride

  • MDL number MFCD13561704
  • Cas Number : 1220021-12-4
  • Molecular Weight : 256.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-260582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-260582
Availability details for
Quantity
  • Availability
  • Add to Cart