ENA497377393 Enamine

6-amino-1,3-dimethyl-5-[2-(pyridin-2-yl)quinoline-4-carbonyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD06328076
  • Cas Number : 945008-17-3
  • Molecular Weight : 387.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-38965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38965
Availability details for
Quantity
  • Availability
  • Add to Cart