ENA497678043 Enamine

2-[(4-chlorophenyl)amino]acetonitrile

  • MDL number MFCD08695331
  • Cas Number : 24889-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135154
Availability details for
Quantity
  • Availability
  • Add to Cart