ENA497678150 Enamine

6-(tert-butoxy)-6-oxohexanoic acid

  • MDL number MFCD24479669
  • Cas Number : 52221-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 10-11
Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182641

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-182641
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart