ENA497678193 Enamine

4-bromothieno[2,3-c]pyridine-2-carboxylic acid

  • MDL number MFCD11846918
  • Cas Number : 1151512-25-2
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-195727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195727
Availability details for
Quantity
  • Availability
  • Add to Cart